missing translation for 'onlineSavingsMsg'
Learn More

Epithelial Membrane Antigen/EMA Ab-2 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Membrane Antigen/EMA Ab-2, Mouse Monoclonal Antibody.

128.00 EUR - 385.00 EUR

Specifications

Antigen Epithelial Membrane Antigen/EMA Ab-2
Clone GP1.4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12621107
View Documents
Epredia
MS-348-P0
Concentrated 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12631107
View Documents
Epredia
MS-348-P1ABX
Concentrated 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12651107
View Documents
Epredia
MS-348-PABX
Concentrated 200 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12641107
View Documents
Epredia
MS-348-P1
Concentrated 500 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12611107
View Documents
Epredia
MS-348-P
Concentrated 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12681107
View Documents
Epredia
MS-348-R7
Prediluted 7 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Epithelial membrane antigen (EMA), is a mucin-like glycoprotein. Antibody to EMA has been shown to be useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in the bone marrow or liver.

Host Species: Mouse

Clone: GP1.4

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: DTRP

Immunogen: Human milk fat globule membranes

Molecular Weight: 265-400 kDa

Positive Control: Breast or Colon CA

Cellular Localization: Cytoplasmic and Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Epithelial Membrane Antigen/EMA Ab-2
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human
IgG1
GP1.4
Monoclonal
Mouse
Oncogenesis
Human milk fat globule membranes
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.