missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epstein Barr Virus BHRF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5117549
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA
Prevents premature death of the host cell during virus production, which would otherwise reduce the amount of progeny virus. Acts as a host B-cell leukemia/lymphoma 2 (Bcl-2) homolog, and interacts with pro-apoptotic proteins to prevent mitochondria permeabilization, release of cytochrome c and subsequent apoptosis of the host cell.Specifications
Epstein Barr Virus BHRF1 | |
Polyclonal | |
Unconjugated | |
BHRF1 | |
Apoptosis regulator BHRF1; EA-R; Early antigen protein R; Nuclear antigen | |
Rabbit | |
Protein G | |
RUO | |
3783706 | |
-20°C or -80°C if preferred | |
Liquid |
Western Blot | |
2.535 mg/mL | |
10mM PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P03182 | |
BHRF1 | |
Recombinant Epstein-Barr virus Apoptosis regulator BHRF1 protein (1-142aa) | |
100 μL | |
Primary | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction