Learn More
Eppendorf™ epTIPS™ Low-retention Pipettor Tips
Increase reproducibility and reduce the loss of expensive reagents in sensitive PCR and real-time PCR applications with these pipettor tips. Eppendorf™ epTIPS™ Low-retention Pipettor Tip Reloads are ideal for cell culture (media) applications and applications with samples that contain detergents.
Specifications
Sterility | Sterile |
---|---|
For Use With (Application) | They satisfy the most stringent demands in medicine, the pharmaceutical and food industries, PCR*, molecular biology and cytotechnology |
Compatibility | Eppendorf™ |
Format | Refill |
Tip Style | Low Retention |
Description
Increase reproducibility and reduce the loss of expensive reagents in sensitive PCR and real-time PCR applications with these pipettor tips. Eppendorf™ epTIPS™ Low-retention Pipettor Tip Reloads are ideal for cell culture (media) applications and applications with samples that contain detergents.
- Ultrahomogenous surface for maximum reproducibility
- Ultrahydrophobic surface for minimal sample loss
- Significantly reduced foam formation during pipetting
- Extreme resistance to chemicals
- Available in PCR clean and Eppendorf Quality
- Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile (sterile and pyrogen-free)
- No coating
Specifications
Sterile | |
Eppendorf™ | |
Low Retention |
They satisfy the most stringent demands in medicine, the pharmaceutical and food industries, PCR*, molecular biology and cytotechnology | |
Refill | |
Eppendorf™ Pipettor |