Learn More
Heidolph™ Erlenmeyer Attachment
Shop All Heidolph Instruments Products
Click to view available options
Holds:
100 mL Erlenmeyer flasks
1000 mL Erlenmeyer flasks
25 mL Erlenmeyer flasks
250 mL Erlenmeyer flasks
50 mL Erlenmeyer flasks
500 mL Erlenmeyer flasks
No. of Positions:
2
4
5
9
14
22
Unit Size:
Each
Description
Attachments for up to 22 Erlenmeyer flasks (25, 50, 100, 250, 500 or 1000 mL)
Specifications
Specifications
| Holds | 250 mL Erlenmeyer flasks |
| Product Type | Erlenmeyer Attachment |
| For Use With (Equipment) | Unimax 1010, Duomax 1030, Polymax 1040 and Promax 1020 |
| No. of Positions | 5 |