Learn More
Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint
Product Code. 17705804
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
200 mL
250 mL
300 mL
500 mL
1000 mL
2000 mL
5 L
Socket Joint:
14/23 NS
19/26 NS
24/29 NS
29/32 NS
45/40 NS
Unit Size:
Each
Pack of 10
Product Code. 17705804
Brand: Lenz Laborglasinstrumente™ 03021922
Only null left
Add to basket
Add to basket
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Certifications/Compliance DIN EN ISO 4797
Description Conical Ground Joint, Inscription Label
Capacity (Metric) 25 mL
Material Borosilicate Glass, Duran Glass
Socket Joint 19/26 NS
Diameter (Metric) Outer 42 mm