missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA586038
This item is not returnable.
View return policy
Description
Rabbit Polyclonal Antibody
This gene encodes a serine hydrolase that belongs to the esterase D family. The encoded enzyme is active toward numerous substrates including O-acetylated sialic acids, and it may be involved in the recycling of sialic acids. This gene is used as a genetic marker for retinoblastoma and Wilson's disease.Specifications
ESD | |
Polyclonal | |
Unconjugated | |
ESD | |
Es10; Es-10; Esd; Esterase 10; esterase D; esterase D/formylglutathione hydrolase; FGH; Methylumbelliferyl-acetate deacetylase; S-formylglutathione hydrolase; Sid 478; sid478; testicular tissue protein Li 66 | |
Rabbit | |
Protein A | |
RUO | |
13885, 2098 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
P10768, Q9R0P3 | |
ESD | |
Recombinant protein within Human ESD aa 1-282 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction