Learn More
mb2 ESD Anti-static Fabric Chair with Normal Lift on Castors
Click to view available options
:
Height: 560mm
Description
- Antistatic work chair
- Gas height adjustment
- Backrest mechanism in height, depth and permanent contact adjustable and in every position lockable
- Conductive plastic parts
- Paddings in polyurethane resin
- Upholstery in antistatic blackfabric
- 5-spoke base in die-cast polished aluminium
- Antistatic double rotating self-braking castors dia. 50 mm
Specifications
Specifications
| Product Type | ESD Chair |
| Color | Gray |
| Adjustable (Height) | Yes |
| Adjustable | Height |
| Autoclavable | Not Autoclavable |
| Back Style | Ergonomic |
| Height (Metric) | 560 mm |
| Seating Height | 420 to 550 mm |
| Upholstery | ESD Fabric |
| Weight (Metric) | 10.70 kg |
| Show More |