missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESRRG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14017-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human ESRRG.
Estrogen-related receptor gamma (ERRG, ERR3, NR3B3) is an orphan member of the nuclear hormone receptor superfamily. ERRG acts as a transcription activator in the absence of a bound ligand. It binds specifically to an estrogen response element and activates reporter genes controlled by estrogen response elements. ERRG also induces the expression of PERM1 in the skeletal muscle.Specifications
ESRRG | |
Polyclonal | |
Unconjugated | |
ESRRG | |
ERR gamma 2, ERR3, ERRG2, ERRgamma, ESRRG, estrogen related receptor gamma, KIAA0832, NR3B3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2104, 26381, 360896 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62508, P62509, P62510 | |
Esrrg | |
ESRRG Fusion Protein Ag5127 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |