Learn More
Estradiol 3-sulfamate, MedChemExpress
MedChemExpress Estradiol 3-sulfamate (BLE 00084; E2MATE; ES-J 995) is a potent, long-acting, and orally active steroid sulfatase inhibitor; inhibits estrone sulfatase with an IC50 of 251 nM and a Ki of 133 nM.
Brand: Medchem Express HY-U00112
This item is not returnable.
View return policy
Description
Estradiol 3-sulfamate (BLE 00084Specifications
Estradiol 3-sulfamate | |
White | |
C18H25NO4S | |
BLE 00084 E2MATE ES-J 995 | |
C[C@@]1([C@H]2O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(OS(N)(=O)=O)=C3)([H])[C@]4([H])CC1 | |
351.46 | |
Research | |
Cancer-programmed cell death |
172377-52-5 | |
Research | |
20 mg | |
DMSO : 62.5 mg/mL (177.83 mM; Need ultrasonic) | |
351.46 | |
99.5% | |
Solid |