missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ether, ULTRA RESI-ANALYZED, J.T. Baker™
Click to view available options
Quantity:
4 L
Specifications
CAS | 60-29-7 |
Quantity | 4 L |
Assay Percent Range | 99.0 % min. |
Molecular Formula | C4H10O |
MDL Number | MFCD00011646 |
Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
SMILES | CCOCC |
IUPAC Name | ethoxyethane |
Molecular Weight (g/mol) | 74.12 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'