missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15448765
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1240278-21-0
Molecular Formula C7H6BrF3N2O2
Molecular Weight (g/mol) 287.036
MDL Number MFCD13193102
InChI Key OVIHBAGNAYIHMQ-UHFFFAOYSA-N
Synonym ethyl 4-bromo-5-trifluoromethyl-1h-pyrazole-3-carboxylate, ethylbromotrifluoromethylpyrazolecarboxylate, ethyl 4-bromo-3-trifluoromethyl-1h-pyrazole-5-carboxylate
PubChem CID 45588074
IUPAC Name ethyl 4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate
SMILES CCOC(=O)C1=NNC(=C1Br)C(F)(F)F

Specifications

Melting Point 131°C to 133°C
Quantity 1 g
Formula Weight 287
Percent Purity 97%
Chemical Name or Material Ethyl 4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'