missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl (4-formyl-2-methoxyphenoxy)acetate

Product Code. 11315569
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11315569

Brand: Alfa Aesar™ H50491.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 51264-70-1
Molecular Formula C12H14O5
Molecular Weight (g/mol) 238.24
MDL Number MFCD01764358
InChI Key SVYWLZTVBMJCGG-UHFFFAOYSA-N
Synonym ethyl 2-4-formyl-2-methoxyphenoxy acetate, ethyl 4-formyl-2-methoxyphenoxy acetate, 4-formyl-2-methoxy-phenoxy-acetic acid ethyl ester, acmc-1ap2o, 4-carbethoxymethoxy-3-methoxybenzaldehyde, 2-methoxy-4-formyl-phenoxyessigsaureathylester, 4-formyl-2-methoxyphenoxy acetic acid ethyl ester, acetic acid, 2-4-formyl-2-methoxyphenoxy-, ethyl ester
PubChem CID 968872
IUPAC Name ethyl 2-(4-formyl-2-methoxyphenoxy)acetate
SMILES CCOC(=O)COC1=C(OC)C=C(C=O)C=C1

Specifications

Quantity 250 mg
Formula Weight 238.24
Chemical Name or Material Ethyl (4-formyl-2-methoxyphenoxy)acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.