missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-methoxyindole-2-carboxylate, 98%

Product Code. p-7047096
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11358536

Brand: Thermo Scientific Alfa Aesar L17596.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4792-58-9 | C12H13NO3 | 219.24 g/mol

It is reactant for preparation of ligands with binding specificity towards the GABA receptor, sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes, anticancer agents,integrase strand-transfer inhibitors (INSTIs),inhibitor of Proliferation of Colon Cancer Cells ,isomeridianin G as GSK-3? inhibitors, HIV-1 integrase inhibitors and inhibitors of mitogen activated protein kinase-activated protein kinase 2 (MK-2).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is reactant for preparation of ligands with binding specificity towards the GABA receptor, sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes, anticancer agents,integrase strand-transfer inhibitors (INSTIs),inhibitor of Proliferation of Colon Cancer Cells ,isomeridianin G as GSK-3? inhibitors, HIV-1 integrase inhibitors and inhibitors of mitogen activated protein kinase-activated protein kinase 2 (MK-2).

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4792-58-9
Molecular Formula C12H13NO3
Molecular Weight (g/mol) 219.24
MDL Number MFCD00047163
InChI Key NPIUAXNFAUGNHP-UHFFFAOYSA-N
PubChem CID 20926
IUPAC Name ethyl 5-methoxy-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=CC(=C2)OC

Specifications

Melting Point 154°C to 157°C
Quantity 1 g
Beilstein 178104
Synonym ethyl 5-methoxyindole-2-carboxylate, 5-methoxy-1h-indole-2-carboxylic acid ethyl ester, ethyl5-methoxyindole-2-carboxylate, ethyl 5-methoxy-2-indolecarboxylate, 1h-indole-2-carboxylic acid, 5-methoxy-, ethyl ester, unii-d63u367gnz, 5-methoxyindole-2-carboxylic acid ethyl ester, methoxy-5 indole carboxylate d'ethyle-2, ethyl 5-methoxyl-1h-indole-2-carboxylate, indole-2-carboxylic acid, 5-methoxy-, ethyl ester
Solubility Information Insoluble in water.
Formula Weight 219.24
Percent Purity 98%
Chemical Name or Material Ethyl 5-methoxyindole-2-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.