missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-bromo-4-chloroquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals

62.91 EUR - 142.71 EUR

Chemical Identifiers

CAS 206257-39-8
Molecular Formula C12H9BrClNO2
Molecular Weight (g/mol) 314.56
MDL Number MFCD00173367
InChI Key YGMLKBFPHVLHGT-UHFFFAOYSA-N
Synonym ethyl 6-bromo-4-chloro-3-quinolinecarboxylate, 4-chloro-6-bromoquinoline-3-carboxylic acid ethyl ester, 3-quinolinecarboxylic acid, 6-bromo-4-chloro-, ethyl ester, 6-bromo-4-chloroquinoline-3-carboxylic acid ethyl ester, ethyl 6-bromo-4-chloro-quinoline-3-carboxylate, ethylbromochloroquinolinecarboxylate, ethyl-6-bromo-4-chloro-quinoline-3-carboxylate, 6-bromo-4-chloro-3-quinolinecarboxylic acid ethyl ester, 6-bromo-4-chloro-quinoline-3-carboxylic acid ethyl esterShow MoreShow Less
PubChem CID 2764138
IUPAC Name ethyl 6-bromo-4-chloroquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN=C2C=CC(Br)=CC2=C1Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15424946
SDS View Documents
Thermo Scientific Alfa Aesar
H59301.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15495568
SDS View Documents
Thermo Scientific Alfa Aesar
H59301.03
1 g
142.71 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ethyl 6-bromo-4-chloroquinoline-3-carboxylate is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-bromo-4-chloroquinoline-3-carboxylate is used as pharmaceutical intermediates

Solubility
Immiscible in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 6-bromo-4-chloroquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.