missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 9-fluorenylideneacetate, 98%, Thermo Scientific™

Product Code. 11449366
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27973-36-0
Molecular Formula C17H14O2
Molecular Weight (g/mol) 250.297
MDL Number MFCD00798562
InChI Key SCHVJDVNASXDFU-UHFFFAOYSA-N
Synonym ethyl 9-fluorenylideneacetate, ethyl 2-9h-fluoren-9-ylidene acetate, ethyl 2-fluoren-9-ylidene acetate, acmc-1cei0, ethyl 9-fluorenylidene acetate, ethyl 9h-fluoren-9-ylideneacetate, ethyl 2-fluoren-9-ylideneethanoate, ethyl 9h-fluoren-9-ylideneacetate #, 9-fluorenylideneacetic acid ethyl ester
PubChem CID 282552
IUPAC Name ethyl 2-fluoren-9-ylideneacetate
SMILES CCOC(=O)C=C1C2=CC=CC=C2C3=CC=CC=C31

Specifications

Melting Point 75°C to 77°C
Quantity 50 g
Formula Weight 250.3
Percent Purity 98%
Chemical Name or Material Ethyl 9-fluorenylideneacetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'