Learn More
Ethyl hydrogen succinate, 95%, Thermo Scientific Chemicals
Product Code. 15471268
Click to view available options
Quantity:
1 g
5 g
25 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1070-34-4
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00053346
InChI Key LOLKAJARZKDJTD-UHFFFAOYSA-N
Synonym monoethyl succinate, ethyl hydrogen succinate, mono-ethyl succinate, butanedioic acid, monoethyl ester, butanedioic acid, 1-ethyl ester, succinic acid monoethyl ester, monoethyl butanedioate, bernsteinsaureathylester, succinic acid monoethyl, succinic acid ethyl ester
PubChem CID 70610
IUPAC Name 4-ethoxy-4-oxobutanoic acid
SMILES CCOC(=O)CCC(=O)O
Specifications
Density 1.141
Boiling Point 248°C
Flash Point 113°C (235°F)
Linear Formula HOOCCH2CH2CO2CH2CH3
Refractive Index 1.432
Quantity 1 g
Formula Weight 146.14
Percent Purity 95%
Chemical Name or Material Ethyl hydrogen succinate