missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenoxyacetate, 99%

Product Code. 11448136
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11448136

Brand: Thermo Scientific Alfa Aesar A14156.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2555-49-9 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2555-49-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00026895
InChI Key MGZFVSUXQXCEHM-UHFFFAOYSA-N
Synonym ethyl phenoxyacetate, acetic acid, phenoxy-, ethyl ester, phenoxyacetic acid ethyl ester, acetic acid, 2-phenoxy-, ethyl ester, phenoxy-acetic acid ethyl ester, phenoxyacetic acid, ethyl ester, ethyl2-phenoxyacetate, pubchem12485, acmc-1ckja, ethylphenoxyacetate
PubChem CID 17365
IUPAC Name ethyl 2-phenoxyacetate
SMILES CCOC(=O)COC1=CC=CC=C1

Specifications

Density 1.108
Boiling Point 251°C to 253°C
Flash Point >110°C (230°F)
Refractive Index 1.5055
Quantity 50 g
Beilstein 1871712
Formula Weight 180.2
Percent Purity 99%
Chemical Name or Material Ethyl phenoxyacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.