Learn More
Masterflex™ Ethyl Vinyl Acetate Microbore Tubing
Product Code. p-8003829
Click to view available options
Diameter (Metric) Inner:
0.25 mm
0.51 mm
0.76 mm
1.02 mm
1.27 mm
1.42 mm
1.52 mm
1.85 mm
Diameter (Metric) Outer:
1.52 mm
2.08 mm
2.29 mm
3.07 mm
3.23 mm
3.35 mm
3.45 mm
3.78 mm
Unit Size:
15m
30m
Product Code. 11977875
Brand: Masterflex™ 9571212
Only null left
Add to basket
Add to basket
Use this tubing for biological testing. Saint Gobain Performance Plastics™ Ethyl Vinyl Acetate Microbore Tubing is biologically stable, nontoxic and non-contaminating.
  • Translucent cross-linked tubing flexible micro-line tubing
  • Virtually 100% elastic memory, returns to its original shape with gentle heating
  • Resin is USP 23 Class VI and National Formulary Vol. 14 specifications
  • Sterilize with autoclave, ethylene oxide, gamma irradiation or dry heat
Specifications
Diameter (Metric) Inner 0.25 mm
Material EVA
Diameter (Metric) Outer 2.08 mm
For Use With Solvent/Hydrocarbon Autoanalysis/Microbore color-coded stopped tubing
For Use With (Application) Solvent/Hydrocarbon Autoanalysis/Microbore color-coded stopped tubing
Min. Operating Temperature (Metric) -50°C
Packaging Quantity 1 Pack
Tubing Size 12
Color Translucent Yellow
Max. Operating Temperature (Metric) 74°C