Learn More
Ethyl vinyl ether, 99%, stab.
Description
Ethyl vinyl ether is used in fragrances, lubricating oil additives and spices preparation. It acts as an intermediate for sulfadiazine. It finds application in pharmaceuticals as anesthetics and analgesics. Further, it is used as a solvent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl vinyl ether is used in fragrances, lubricating oil additives and spices preparation. It acts as an intermediate for sulfadiazine. It finds application in pharmaceuticals as anesthetics and analgesics. Further, it is used as a solvent in organic synthesis.
Solubility
Miscible with common organic solvents. Slightly miscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents, oxygen and acids. Store in a cool place.
Specifications
Specifications
| Melting Point | -115°C |
| Density | 0.754 |
| Boiling Point | 33°C to 34°C |
| Flash Point | −45°C (−49°F) |
| Odor | Ether-like |
| Linear Formula | CH2=CHOCH2CH3 |
| Refractive Index | 1.3765 |
| Quantity | 500 mL |
| UN Number | UN1302 |
| Beilstein | 605351 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.