missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol-bis(Succinic acid N-hydroxysuccinimide ester), MP Biomedicals™

Product Code. 11455421 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11455421

Brand: MP Biomedicals 0215798291

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A homobifunctional, cleavable cross-linking reagent; It is typically, coupled to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5 to 8.5). The linkage provides a 12-atom spacer

TRUSTED_SUSTAINABILITY

Specifications

CAS 70539-42-3
Color White
Molecular Formula C18H20N2O12
MDL Number MFCD00049057
Quantity 1 g
Synonym ethylene glycol-bis succinic acid n-hydroxysuccinimide ester, ethylene glycol bis succinimidyl succinate, bicl110, 1,2-ethanediyl bis 4-2,5-dioxo-1-pyrrolidinyl oxy-4-oxobutanoate, ethylene glycol-bis succinimidyl succinate, bis 2,5-dioxopyrrolidin-1-yl o,o'-ethane-1,2-diyl disuccinate, 2,5-dioxopyrrolidin-1-yl 1-2-4-2,5-dioxopyrrolidin-1-yl oxy-4-oxobutanoyl oxy ethyl butanedioate, egs, egbss, ethylene glycolylbis succinimidyl succinate
InChI Key QLHLYJHNOCILIT-UHFFFAOYSA-N
SMILES O=C(CCC(=O)ON1C(=O)CCC1=O)OCCOC(=O)CCC(=O)ON1C(=O)CCC1=O
IUPAC Name 2,5-dioxopyrrolidin-1-yl 1-[2-({4-[(2,5-dioxopyrrolidin-1-yl)oxy]-4-oxobutanoyl}oxy)ethyl] butanedioate
Molecular Weight (g/mol) 456.36
PubChem CID 123663
Formula Weight 456.4
Physical Form Powder
Chemical Name or Material Ethylene glycol-bis(Succinic acid N-hydroxysuccinimide ester)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.