Learn More
Ethylenebis(isothiouronium bromide), 98%, Thermo Scientific™
Product Code. 11326996
Click to view available options
Quantity:
5 g
25 g
100 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6943-65-3
Molecular Formula C4H12Br2N4S2
Molecular Weight (g/mol) 340.096
MDL Number MFCD00035055
InChI Key VABTXEYLYWGJRB-UHFFFAOYSA-N
Synonym ethylenediisothiouronium dibromide, ethylene diisothiouronium dibromide, ethylenediisothiourea dihydrobromide, ethylene diisothiourea dihydrobromide, pseudourea, 2,2-ethylenedithiodi-, dihydrobromide, 2,2'-ethylenebisisothiouronium dibromide, carbamimidothioic acid, 1,2-ethanediyl ester, dihydrobromide, ethylenebis isothiouronium bromide, 2,2'-ethylene-bis-2-thiopseudourea , dihydrobromide, pseudourea, 2,2'-ethylenebis 2-thio-, dihydrobromide
PubChem CID 81370
IUPAC Name 2-carbamimidoylsulfanylethyl carbamimidothioate;dihydrobromide
SMILES C(CSC(=N)N)SC(=N)N.Br.Br
Specifications
Melting Point 224°C to 226°C
Quantity 25 g
Beilstein 3914616
Sensitivity Hygroscopic
Formula Weight 340.09
Percent Purity 98%
Chemical Name or Material Ethylenebis(isothiouronium bromide)