missing translation for 'onlineSavingsMsg'
Learn More

Europium rod, 9.52 mm (0.375in) dia, 99.9% (metals basis)

Product Code. p-7059411
Click to view available options
Quantity:
10 mm
50 mm
Unit Size:
10mm
50mm
This item is not returnable. View return policy

Product Code. 11465300

Brand: Thermo Scientific Alfa Aesar 044545.HF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-53-1 | Eu | 151.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in mineral oil
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-53-1
Molecular Formula Eu
Molecular Weight (g/mol) 151.96
MDL Number MFCD00010992
InChI Key OGPBJKLSAFTDLK-UHFFFAOYSA-N
Synonym europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil
PubChem CID 23981
ChEBI CHEBI:32999
IUPAC Name europium
SMILES [Eu]

Specifications

Color Gray-Silver
Quantity 10 mm
Odor Odorless
Weight ≈0.37g/mm
Form Cast Rod
UN Number UN3208
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Europium rod, ≈9.52 mm (0.375 in.) dia.
compliance-icons
Product Identifier
  • Europium rod
Signal Word
  • Danger
Hazard Category
  • Pyrophoric liquid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Europium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.