Learn More
IKA™ EUROSTAR 60 Digital Overhead Stirrer
Universally used strong laboratory stirrer for quantities up to 40 L (H2O). It automatically adjusts the speed through microprocessor controlled technology within the speed range.
Brand: IKA™ 0004446003
1594.75 EUR valid until 2024-12-31
Use promo code "21913" to get your promotional price.
This item is not returnable.
View return policy
Description
- Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions
- Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically
- Guarantees a constant speed even with changes in viscosities of the sample
- Digital speed display
- Infinitely adjustable speed
- Push-through agitator shafts
- Overload protection
- Short-term overload operation
- Slim casing
- Quiet operation
- Error code display
Specifications
30 to 2000 rpm | |
7 Segment LED | |
176 w | |
50/60 Hz | |
Alu-Cast Coating/Thermoplastic Polymer | |
5°C to 40°C |
40 L | |
100/115/230 V | |
208 x 86 x 248 mm | |
CH-Style | |
Brushless DC | |
4.4 kg |