Learn More
Thermo Scientific™ Eutech TN-100 Handheld Infrared Turbidity Meter Kit
Waterproof, portable, infrared Turbidity meter wtih extended range up to 2000 NTU, 0.01 resolution
Brand: Thermo Scientific™ 01X357301
920.38 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Includes: Set of 4 calibration standards (800, 100, 20.0 & 0.02 NTU)
Description
Waterproof, portable, infrared Turbidity meter wtih extended range up to 2000 NTU, 0.01 resolution
- Waterproof
- Set of 7 sample vials
- Lint-free cloth
- Silicon oil
- Batteries
- Hard carrying case
Specifications
Infrared Turbidity Meter Kit | |
U.S. EPA-approved non-formazin standards | |
0.01 | |
ISO 7027 compliant |
Portable, Waterproof | |
Waterproof TN-100 handheld turbidimeter with set of 4 calibration standards (800, 100, 20.0 & 0.02 NTU), set of 7 sample vials, lint-free cloth, silicone oil, batteries, and hard carrying case. | |
Up to 2000 NTU |
Product Suggestions
Customers who viewed this item also viewed
Water analysis
File size: 7.3 MB
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
Product Content Correction
Product Title
Thermo Scientific™ Eutech TN-100 Handheld Infrared Turbidity Meter Kit >
Spot an opportunity for improvement?Share a Content Correction