Learn More
Eutech™ Conductivity Calibration Sachets
Get quick and accurate conductivity measurements in the field, laboratories, and schools.
Brand: Thermo Scientific Eutech ECCON2764BS
This item is not returnable.
View return policy
Description
Get quick, accurate, and convenient conductivity measurements with NIST_traceable Eutech™ Conductivity Calibration Sachets. Simply insert electrode into sachet pack, calibrate, rinse and discard sachet – no separate container needed.
Features:
- Conductivity Standards Sachet -20 ml
- NIST Traceable
Specifications
2764 μS/cm KCl Conductivity Sachets | |
Calibration Solution | |
Conductivity Standard | |
Field, School, Laboratory |