Learn More
Lenz Laborglasinstrumente™ Evaporating Flask with Conical Ground Joint
Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ Evaporating Flasks with Conical Ground Joints can be used to make solutions or to hold, contain, collect or measure chemicals and are manufactured from durable DURAN™ glass.
Brand:  Lenz Laborglasinstrumente™ 03042970
Product Code. 10212151
  • 63.70 EUR / Each
In Stock
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Description
Description
  • With center neck and conical joint
  • Pear-shaped with inscription label
  • Available in a range of capacities
Specifications
Specifications
1000 mL
Rotary Evaporators
126 mm
Borosilicate Glass, Duran Glass
Center