Learn More
SI Analytics™ Exchange Unit
SI Analytics™ Exchange Unit is a unit that includes an integrated chip for reagent dat, tubes, a titration tip, and more.
Brand: SI Analytics™ 285220320
This item is not returnable.
View return policy
Description
- Includes integrated chip for reagent data
- Comes with a brown glass bottle for titrant
- GL 45
- S 40 bottle adapter
- Includes tubes, drip tubes, and titration tip
Specifications
Brown glass bottle for titrant | |
Exchange Unit | |
20 mL |
GL 45 | |
Integrated chip for reagent data |