Learn More
BINDER™ Expanded Calibration Certificate for Temperature and Humidity
Product Code. 30238319 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KBF PRO Series
KBF Series
Unit Size:
Each
Product Code. 30238319
Brand: BINDER™ 80122576
Certificate that includes an expanded range of temperature and humidity measurements.
Certificate for extending the measurement in the center of the chamber to include another test value.
Specifications
For Use With (Equipment) KBF PRO Series
Duration 1 Day
Product Type Calibration Certificate