Learn More
Cytiva Exposure Cassette for Unmounted Screen
Protects the screen from light during exposure to the sample.
474.00 EUR - 708.00 EUR
Specifications
For Use With (Equipment) | Unmounted Screen |
---|---|
For Use With (Application) | Light exposure |
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
15824182
|
Cytiva
29175523 |
20 x 25 cm |
474.00 EUR
Each |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15834182
|
Cytiva
29175524 |
35 x 43 cm |
708.00 EUR
Each |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For use with unmounted screens (not included).
- Protects your screen from light during exposure.
Specifications
Unmounted Screen | |
Light exposure |