missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FABP1 Monoclonal Antibody (5B5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA121432
This item is not returnable.
View return policy
Description
MA1-21432 detects Fatty Acid Binding Protein in Human samples.
FABP1 encodes the fatty acid binding protein found in liver. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 and FABP6 (the ileal fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.
Specifications
| FABP1 | |
| Monoclonal | |
| 7.8 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P07148 | |
| FABP1 | |
| Human heart FABP. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Affinity Purification, ELISA, Immunohistochemistry | |
| 5B5 | |
| Unconjugated | |
| FABP1 | |
| 14 kDa selenium-binding protein; FABP1; FABPL; Fabplg; fatty acid binding protein 1; fatty acid binding protein 1, liver; fatty acid binding protein liver; fatty acid-binding protein 1; Fatty acid-binding protein, liver; L-FABP; Liver-type fatty acid-binding protein; p14; SCP; SCP.; Squalene- and sterol-carrier protein; Z-protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 2168 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction