Learn More
Memmert™ Factory Calibration (3 Temperatures) for UF75plus
Calibration at Memmert factory for three temperature value.
Brand:  Memmert™ UF75plusD00128
Product Code. 17579695
  • 356.00 EUR / Each
Description
  • For three temperature values: 100°C, 160°C, and 220°C
Specifications
Specifications
UF75plus Oven
Measuring point chamber center
Certificate for 3 temperatures
Standard Works Calibration Certificate