missing translation for 'onlineSavingsMsg'
Learn More

Farnesylpyrophosphate, 98%, MP Biomedicals™

Product Code. 11410092
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 13058-04-3
Assay Percent Range 0.98
Molecular Formula C15H37N3O7P2
Quantity 1 mg
Synonym farnesylpyrophosphate, farnesyl pyrophosphate triammonium salt, farnesyl pyrophosphate ammonium salt, 3,7,11-trimethyl-2,6,10-dodecatrien-1-yl pyrophosphate ammonium salt, fpp, 3,7,11-trimethyl-2,6,10-dodecatrien-1-ol pyrophosphate, triamine farnesol pyrophosphate, farnesyl diphosphate triamine, farnesyl pyrophosphate ammonium salt, in methanol
InChI Key GSWRJNPTNZUHOV-VLCVYXDNSA-N
SMILES CC(=CCCC(=CCCC(=CCOP(=O)(O)OP(=O)(O)O)C)C)C.N.N.N
IUPAC Name azane;phosphono [(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl] hydrogen phosphate
Molecular Weight (g/mol) 433.423
PubChem CID 71299715
Formula Weight 442.5
Physical Form Solution
Chemical Name or Material Farnesylpyrophosphate, For TLC analysis
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'