Learn More
Fc gamma RIIA/CD32a Antibody (IV.3) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20159-0.025mg
 View more versions of this product
Product Code. 18650294
  • 267.00 EUR / 0.02mg
Description
Fc gamma RIIA/CD32a Monoclonal antibody specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Inhibition Activity, Neutralization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fc gamma RIIA/CD32a
Monoclonal
1 mg/mL
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Inhibition Assays
IV.3
Unconjugated
CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a
The original antibody was generated against the FcR of K562 cells.
0.025 mg
Immunology
2212
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions