Learn More
Fehling'S Solution No. 2, Contains Sodium Hydroxide To B.P. 1988, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
Specifications
Specifications
| Color | Colorless |
| Packaging | HDPE plastic bottle |
| Quantity | 1 L |
| Physical Form | Liquid |