Learn More
Saint-Gobain FEP Tubing
Brand:  Saint-Gobain C20080X04010
Product Code. 11757335
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
FDA compliant (food), USP Class VI, manufactured according to GLP
0.8 mm
1.6 mm
0.4 mm
-270°C
Sterile
Semirigid
205°C
Autoclavable
FEP
10 m
Transfer fluid pressure or high temperature in the chemical, pharmaceutical, mechanical, electrical, automobile
1 Each
Translucent
34 bar