Learn More
Thermo Scientific™ NoLimits 15000 bp DNA Fragment
GREENER_CHOICE
Product Code. 10519120 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. 10519120
Brand: Thermo Scientific™ SM1531
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology.
Plasmid DNA is digested with Thermo Scientific Restriction Enzymes, and the individual DNA fragments are chromatography-purified from the digestion mixture. As a result, we offer exceptionally pure DNA fragments that are free of any truncated or degraded molecules. Our custom DNA fragments are the best choice for applications, ranging from electrophoresis (both gel and capillary) to HPLC and beyond.
Highlights
  • Chromatography-purified individual DNA fragments
  • Broad range: 10 to 20,000 bp
  • Sharp peaks during capillary electrophoresis, sharp bands during gel electrophoresis
  • Precise DNA concentration 
TRUSTED_SUSTAINABILITY
Specifications
Gel Type Agarose
Product Type DNA Fragment
Ready to Load No
Size Range 10 to 20000 bp
Gel Compatibility Agarose Gels
Green Features Sustainable packaging
Product Line NoLimits
Quantity 10 μg