Learn More
Thermo Scientific™ Rapid DNA Ligation Kit
Enable fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature.
Brand:  Thermo Scientific™ K1422
147.68 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
 View more versions of this product
Product Code. 10775841
  • 212.00 EUR / 50 reactions
Edge
Description
Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature.
The kit contains T4 DNA ligase and a specially-formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation. The ligation reaction mixture can be used directly for bacterial transformation with the TransformAid Bacterial Transformation Kit or with other conventional transformation procedures.
The kit contains T4 DNA ligase and a specially formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation. The ligation reaction mixture can be used directly for bacterial transformation with the TransformAid Bacterial Transformation Kit or with other conventional transformation procedures.
  • Fast—ligation of either sticky- or blunt-end DNA in only 5 minutes
  • Convenient—reaction mixture can be used directly for bacterial transformation
Recommended for:
Routine cloning experiments; Blunt-end cloning; Library construction; TA cloning.
Note:
Lyophilized bacterial strains GM2163 (†M0099) and JM107 (†M0109) can be provided free of charge upon request, with any purchase. Prior to electroporation it is necessary to inactivate T4 DNA ligase (1) by chloroform extraction.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5X Rapid Ligation Buffer
DNA Ligase
Rapid DNA Ligation Kit
50 Reactions
Documents
Manuals
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2822089 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1422
Lot Number2825198 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1422
Lot Number2792572 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1422
Lot Number2841211 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1422
Lot Number2866370 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberK1422
Product Title
Thermo Scientific™ Rapid DNA Ligation Kit > 50 reactions