missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Pyrophosphate, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| CAS | 10058-44-3 |
| Quantity | 100 g |
| Molecular Formula | Fe4O21P6 |
| MDL Number | MFCD00016091 |
| Synonym | ferric pyrophosphate, iron iii pyrophosphate, iron pyrophosphate, iron 3+ pyrophosphate, tetrairon tris pyrophosphate, ccris 1900, hsdb 454, diphosphoric acid, iron 3+ salt 3:4, pyrophosphoric acid, iron 3+ salt 3:4, sunactive fe |
| InChI Key | CADNYOZXMIKYPR-UHFFFAOYSA-B |
| SMILES | [Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O |
| IUPAC Name | tetrairon(3+) tri(phosphonatooxy)phosphonate |
| Molecular Weight (g/mol) | 745.20 |
| PubChem CID | 24877 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction