missing translation for 'onlineSavingsMsg'
Learn More

Ferritin, Equine, MP Biomedicals™

Product Code. 11446922
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Plays a key role in iron metabolism. Ferritin, Equine, MP Biomedicals is a ubiquitous iron storage protein with a shell of 24 subunits (apoferritin). It serves as an intracellular iron reserve (particularly in spleen, liver, intestinal mucosa, bone marrow

A shell of 24 protein subunits (apoferritin) and a core of Fe3+ ions with a peptide molecular weight of 440 kDa. The ferritin-like superfamily of iron-storage proteins is virtually ubiquitous in life on earth.

  • Studies have shown that ferritin iron incorporation is mediated by a ferroxidase activity associated with ferritin H subunits and a nucleation center associated with ferritin L subunits.
  • Release of iron from ferritin has an essential role in iron-dependent lipid peroxidation and may contribute to free radical-induced cell damage in vivo.
  • Therefore, by binding iron, ferritin may function as an antioxidant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9007-73-2
Molecular Formula C16H33O2S-
Molecular Weight (g/mol) 289.498
InChI Key HBTBNXFVJYRYGI-UHFFFAOYSA-M
Synonym 1exw, hsf
PubChem CID 6419224
IUPAC Name hexadecane-1-sulfinate
SMILES CCCCCCCCCCCCCCCCS(=O)[O-]

Specifications

Quantity 100 mg
Solubility Information Soluble in 0.15M sodium chloride, distilled water or dilute buffer.
Formula Weight ∼450,000 (Lit.)
Physical Form Viscous Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'