missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin, Light Chain Mouse, Clone: FTL/1386, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2512-MSM1-P0
This item is not returnable.
View return policy
Specifications
Ferritin, Light Chain | |
Cytoplasmic | |
FTL/1386 | |
Unconjugated | |
NP_000137 | |
Mouse | |
19-25kDa | |
Protein A/G Purified | |
RUO | |
2512 | |
IgG2b |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
FTL | |
Recombinant human FTL protein fragment (aa 38-165) | |
HepG2, HeLa, HL-60 or 293T cells. Pancreas, Liver, Cerebellum or Testis. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction