missing translation for 'onlineSavingsMsg'
Learn More

Ferritin, Light Chain Mouse, Clone: FTL/1386, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 2512-MSM1-P1

 View more versions of this product

Product Code. 15960058

  • 458.00 EUR / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Ferritin, Light Chain
Cytoplasmic
FTL/1386
Unconjugated
NP_000137
Mouse
19-25kDa
Protein A/G Purified
RUO
2512
IgG2b
Western Blot
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
FTL
Recombinant human FTL protein fragment (aa 38-165)
HepG2, HeLa, HL-60 or 293T cells. Pancreas, Liver, Cerebellum or Testis.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers