Learn More
Trajan Scientific and Medical™ Ferrule SilTite™ Metal Kit For Agilent Split/Splitless Injectors
Product Code. 12987091
Click to view available options
For Use With (Equipment):
Agilent Split/Splitless Injector
Unit Size:
1 set
Product Code. 12987091
Brand: Trajan Scientific and Medical™ 073272
Only null left
Add to basket
Add to basket
Delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Specifically designed for connecting capillary columns
  • Connects guard columns to capillary columns
  • Inert and precise glass lined bore
  • Low dead volume
  • Suitable for Agilent GCs
Specifications
Type Ferrule SilTite Metal Kit
For Use With (Equipment) Agilent Split/Splitless Injector
Quantity 1 Each
Diameter (Metric) Inside 0.45 to 0.53 mm