Learn More
Trajan Scientific and Medical™ Ferrule SilTite™ Metal Kit For Agilent Split/Splitless Injectors
Click to view available options
For Use With (Equipment):
Agilent Split/Splitless Injector
Unit Size:
1 set
Description
- Specifically designed for connecting capillary columns
- Connects guard columns to capillary columns
- Inert and precise glass lined bore
- Low dead volume
- Suitable for Agilent GCs
Specifications
Specifications
| Type | Ferrule SilTite Metal Kit |
| For Use With (Equipment) | Agilent Split/Splitless Injector |
| Quantity | 1 Each |
| Diameter (Metric) Inside | 0.45 to 0.53 mm |