Learn More
FGF21 Monoclonal Antibody (OTI9A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA502850S
Encompass_Preferred
 View more versions of this product
Product Code. 17587911
  • 242.00 EUR / 30µL
Explore more special offers
Description
Description
FGF21 Monoclonal antibody specifically detects FGF21 in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications
Specifications
FGF21
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P11086
Mouse
Affinity chromatography
RUO
5409
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Flow Cytometry, Western Blot
OTI9A7
Unconjugated
Pnmt
FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196
Full length human recombinant protein of human PNMT produced in HEK293T cell.
30 μL
Primary
Human
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
FGF21 Monoclonal Antibody (OTI9A7), TrueMAB™, OriGene