Learn More
Velp Scientifica™ Fine and Coarse Stator
Velp Scientifica™ Fine and Coarse Stator can be reconfigured with a range of OV5 Homogenizer Shafts and Rotors.
Brand: Velp Scientifica™ A00000052
This item is not returnable.
View return policy
Description
Velp Scientifica™ Fine and Coarse Stator can be reconfigured with a range of OV5 Homogenizer Shafts and Rotors.
- Can be romoved and reconfigured
- Interchangable
- User-friendly
- Strong and durable
Specifications
Coarse Stator | |
Stator for homogenizer | |
VS4C |
40 mm | |
OV5 Homogenizer | |
1 Ea. |