Learn More
Thermo Scientific™ Finnpipette™ F1 Fixed Volume Single Channel Pipettes
Choose these pipettes for set protocols. They deliver a light, smooth pipetting action and an adjustable finger rest to maximize your comfort.
Specifications
Description | Finnpipette F1 Fixed Volume |
---|---|
Number of Channels | 1 |
Type | Fixed Volume Pipette |
Product Code | Brand | Description | Volume (Metric) | Compatible Tips | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) | Compatible Tips | Price | Quantity & Availability | |||
15541905
|
Thermo Scientific™
4651020N |
Finnpipette F1 Fixed Volume | 10 μL | Finntip 250, Filter 10, Flex 200, Flex Filter 30 |
N/A
|
N/A | |||
15551905
|
Thermo Scientific™
4651030N |
Finnpipette F1 Fixed Volume | 25 μL | Finntip 250, Filter 30, Flex 200, Flex Filter 30 |
N/A
|
N/A | |||
15561905
|
Thermo Scientific™
4651040N |
Finnpipette F1 Fixed Volume | 50 μL | Finntip 250, Filter 100, Flex 200, Flex Filter 100 |
N/A
|
N/A | |||
15571905
|
Thermo Scientific™
4651050N |
Finnpipette F1 Fixed Volume | 100 μL | Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide |
N/A
|
N/A | |||
15581905
|
Thermo Scientific™
4651060N |
Finnpipette F1 Fixed Volume | 250 μL | Finntip 1000, Filter 1000, Flex 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext |
N/A
|
N/A | |||
15591905
|
Thermo Scientific™
4651070N |
Finnpipette F1 Fixed Volume | 500 μL | Finntip 1000, Filter 1000, Flex 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext |
N/A
|
N/A | |||
15501915
|
Thermo Scientific™
4651080N |
Finnpipette F1 Fixed Volume | 1000 μL | Finntip 1000, Filter 1000, Flex 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext |
N/A
|
N/A | |||
15511915
|
Thermo Scientific™
4651090N |
Finnpipette F1 Fixed Volume | 2000 μL | Finntip 5mL, Filter 5mL |
N/A
|
N/A | |||
15521915
|
Thermo Scientific™
4651100N |
Finnpipette F1 Fixed Volume | 3000 μL | Finntip 5mL, Filter 5mL |
N/A
|
N/A | |||
15531915
|
Thermo Scientific™
4651110N |
Finnpipette F1 Fixed Volume | 5000 μL | Finntip 5mL, Filter 5mL |
N/A
|
N/A | |||
15541915
|
Thermo Scientific™
4651120N |
Finnpipette F1 Fixed Volume | 10,000 μL | Finntip 10mL, Filter 10mL, Flex 10mL Ext, Flex Filter 10mL Ext |
N/A
|
N/A | |||
15551915
|
Thermo Scientific™
4651130N |
Finnpipette F1 Fixed Volume | 20 μL | Finntip 250, Filter 20, Flex 200, Flex Filter 30 |
N/A
|
N/A | |||
15561915
|
Thermo Scientific™
4651140N |
Finnpipette F1 Fixed Volume | 200 μL | Finntip 250, Filter 200, Flex 200, Flex Filter 200, 200 Ext, Filter 200 Ext, 250 Wide |
N/A
|
N/A | |||
Description
Great for set protocols and QA environments, Thermo Scientific™ Finnpipette™ F1 Fixed Volume Pipettes deliver a light, smooth pipetting action and an adjustable finger rest to maximize your comfort.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >
Specifications
Finnpipette F1 Fixed Volume | |
1 | |
Fixed Volume Pipette |