Learn More
Thermo Scientific™ Finntip™ Stepper Pipette Tips
GREENER_CHOICE
Product Code. 10762111
Click to view available options
Volume (Metric):
500 μL
1250 μL
2.5 mL
5 mL
12.5 mL
25 mL
50 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 25
Pack of 50
Product Code. 10762111
Brand: Thermo Scientific™ 9404200
Only null left
Add to basket
Add to basket
Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
  • Nonsterile tips are bulk packaged; sterile tips are individually wrapped
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 5 mL
Volume (Metric) Dispensing 100, 200, 300, 400, 500 μL
Filtered Non-filtered
Sterility Non-sterile
Certifications/Compliance ISO 9001, ISO 14001
Format Bulk
For Use With (Equipment) Thermo Scientific FinnPipette Stepper
Product Line FinnTip