Learn More
3M™ First Base™ + Bump Cap
Click to view available options
Product Type:
Micro Peak
Reduced Peak
Standard Peak
Unit Size:
Pack of 20
Color:
Black
Gray
Navy Blue
Royal Blue
Description
- Provides industrial bump protection to EN812.
- Flexible shell made with ABS plastic, can be removed for cleaning.
- Airflow channels for ventilation.
- Absorbent terry toweling sweatband.
- Available in different peak lengths for a variety of applications.
- Adjustable elastic strap for a personal fit.
- Optional logo customization areas.
- Available in different peak lengths for a variety of applications.
- Adjustable elastic strap for a personal fit.
- Belt clip accessory available.
Specifications
Specifications
| Product Type | Reduced Peak |
| Material | UV-Stabilized ABS |