missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ EZ-Run™ Protein Marker
Mixture of seven purified proteins optimized to produce uniform, compact bands when analyzed by SDS-PAGE and stained with Coomassie™ Blue
Brand: Fisher BioReagents™ BP3600-1
This item is not returnable.
View return policy
Description
- Supplied in gel loading buffer for direct application to SDS-PAGE gels
- Ideal for molecular weight determination of polypeptides analyzed in various gel systems
Specifications
Protein Molecular Weight Marker/Ladder | |
Supplied in gel-loading buffer | |
<51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet | |
14.4 to 116 kd | |
200 Loadings |
Liquid | |
Monitoring protein separation during SDS-PAGE | |
2 x 500 μL, 200 Loadings | |
−20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction