Learn More
Fisherbrand™ Replacement Paddle for accumet™ Probe
For use with Fisherbrand accumet benchtop stirring Probe (Cat. No. 13-620-BSP).
Brand: Fisherbrand™ 13-620-RP
This item is not returnable.
View return policy
Description
- Replacement paddle with stirring rod
- Fisher Scientific accumet benchtop stirring probe is used with XL series Benchtop Meters (Models XL150, 200, 250, 500, and 600) and AB series (Models AB150, 200 and 250)
Order Info
Compatible with the following models: XL150, XL200, XL250, XL500, XL600, AB150, AB250, AB200
Services
One year
Specifications
Replacement paddle |