Learn More
Fisherbrand™ Vortex Mixer Accessories: Head and Head Cover
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Fisherbrand Vortex mixer
Unit Size:
Each
Description
For use with Fisherbrand vortex mixers.
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
- 3 in. Head permits easy-touch mixing of beakers and flasks
- Head Cover slides over top of head—ideal for odd-shaped containers
- Head With Rubber Cover is designed for mixing irregular shaped objects
Specifications
Specifications
| Product Type | Head with Cover |
| Diameter (Metric) | 76 mm |
| For Use With (Equipment) | Fisherbrand Vortex mixer |
| Includes | Head, Rubber cover |